missing translation for 'onlineSavingsMsg'
Learn More

U 73343, Tocris Bioscience™

Product Code. 30001464
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Analog of U 73122 (Cat. No. 1268)

Analog of U 73122 ; can be used as a negative control. Inhibits Panx1 currents in HEK cells. Also inhibits vasopressin- and GTPγS-induced Ca2+ influx in hepatocytes.

Active Analog also available.

Chemical Identifiers

CAS 142878-12-4
Molecular Formula C29H42N2O3
Molecular Weight (g/mol) 466.67
MDL Number MFCD00211221
InChI Key CJHWFIUASFBCKN-ZRJUGLEFSA-N
Synonym unii-s2c4j8704c, 1-6-17beta-3-methoxyestra-1,3,5 10-triene-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl-amino hexyl-2,5-pyrrolidine-dione, 1/6/2017, a-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17?-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidine-dione, 1-6-8r,9s,13s,14s,17s-3-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta a phenanthren-17-yl amino hexyl pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl
PubChem CID 114825
IUPAC Name 1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)pyrrolidine-2,5-dione
SMILES COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)CCC1=O

Specifications

Quantity 10 mg
Formula Weight 466.66
Percent Purity >98%
Chemical Name or Material U 73343
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'