Learn More
Labcon U-Pette™ Blood Transfer Dispensers
These blood transfer dispensers are for use with point of care testing instruments.
Brand: Labcon 7626-500-000-9
535.57 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
U-Pette™ Blood Dispenser for Point of Care Test Cartridges dispenses blood drops easily with no need to open collection tubes.
- Dispenses one or more drops of blood
- Fits standard glass or plastic blood tubes with rubber stoppers
- Includes Stainless Steel Cannula
- Not Autoclavable
Specifications
Blood Droplet Dispenser | |
Single-use | |
Point of Care Testing Instruments | |
500 Pk. |