missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ UAS-bla HEK 293T Master Cell Line

The UAS-bla HEK 293T Master Cell Line consists of HEK 293T cells stably transfected with a UAS-bla fusion construct.

Brand:  Thermo Scientific™ K1711

Product Code. 10494035

  • 11430.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The UAS-bla HEK 293T Master Cell Line consists of HEK 293T cells stably transfected with a UAS-bla fusion construct. This cell line can be used to build cell-based assays by transfecting with GAL4 fusion constructs. Activation of the GAL4 fusion results in binding to the UAS-bla and subsequent expression of beta lactamase. Expression of beta lactamase is monitored using a FRET-based assay.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dividing Cells
Fluorescence, Fluorescent
UAS-bla HEK 293T Master Cell Line, 2 x 106cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, Fluorescent Detection Method, BLA (Beta-Lactamase) Reporter, Adherent Growth Properties, Liquid Nitrogen Storage
Cellular imaging, pharma and biopharma, cell analysis, cellular imaging, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, target and lead identification and validation, nuclear receptor biology, cell-based nuclear receptor reporter assays
2 x 106 Cells
2 x 106 Cells, 2 x 106 cells
293T (HEK)
Cell Line
Shipping: Dry Ice
Storage: Liquid Nitrogen, Liquid Nitrogen
293T (HEK)
Adherent Cell Culture
HEK 293T cells stably transfected with a UAS-bla fusion construct
GeneBLAzer™
HEK 293T Cells
BLA (Beta-Lactamase)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.