missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90
BIOPHARMAGREENER_CHOICE

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3045800135308

 View more versions of this product

Product Code. 15924506

  • 2055.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly:  drawers remain open in any position, interior of drawers completely visible – access to all containers
  • Safe: drawers fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: drawers lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Services

Five years

Specifications

Specifications

Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
502 mm
382 x 771 x 502 mm
Sliding
600 mm
UB90.060.089.050.S
Powder-coated plain
893 mm
Light Gray
Safety storage under bench cabinet
1 x 17.5 liter drawer (sheet steel powder-coated)
382 mm
502 x 893 x 600 mm
Safe and approved storage of flammable hazardous substances in working areas
502 mm
2
148 kg
771 mm
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers