missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3047300135295
1333.11 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
For indoor use only
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- User-friendly: drawers remain open in any position, interior of drawers completely visible – access to all containers
- Safe: drawers fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
- No unauthorised use: drawers lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Services
Five years
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
502 mm | |
382 x 470 x 502 mm | |
Sliding | |
600 mm | |
UB90.060.059.050.S | |
Powder-coated plain | |
593 mm | |
Light Gray | |
Safety storage under bench cabinet |
1 x drawer (sheet steel powder-coated) | |
382 mm | |
502 x 593 x 600 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
502 mm | |
1 | |
111 kg | |
470 mm | |
Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction