missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ UB-S-90 Underbench Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances under work benches and fume cupboards.
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Locking |
Description
- Cabinet makes perfect use of the space available under the fume cupboard
- Available in a range of sizes and configurations to enable maximum flexibility in adapting to user requirements and structural conditions
- SOFT CLOSE system – no matter how large the force while sliding the drawer, thanks to spring mechanisms the drawer closes softly and quietly
- Maximized interior height for high storage capacity
- Problem-free connection to supply and disposal pipes
- No unauthorized use – doors and drawers are lockable with an integrated locking state indicator (red/green) to signal the locking status
- Fire resistance of 90 minutes
- In accordance with EN 14470-1 and TRGS 510
- Available with integrated drawers, second level drawers and bottom collecting sumps
- Safe – drawers fitted with earthing wire including clamp as standard, seal welded sump, and self-locking drawers in the event of fire
- Robust construction and longevity – body made of powder-coated sheet steel
- User-friendly and safe – doors remain open in any position, interior of drawers completely visible
- Ventilation – integrated air ducts ready for connection (DN 50) to a technical exhaust system
Specifications
EN 14470-1, TRGS 510 | |
Steel | |
Locking | |
Powder-coated |
Light Gray | |
Safety storage cabinet | |
Storage of flammable and hazardous substances |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title