Learn More
SI Analytics™ Ubbelohde Viscometer (DIN)
Ubbelohde viscometers are calibrated for manual measurements
Brand: SI Analytics™ 285400115
Description
- Calibrated for manual measurement
- DIN
- Type No. 501 31
- Capillary No. IIIa
- Constant K = 5
Specifications
Ubbelohde Viscometer |