missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBC13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10243-1-AP-150UL
This item is not returnable.
View return policy
Description
Anti-Ubc13 is a monoclonal antibody specific to the approximately19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.Specifications
UBC13 | |
Polyclonal | |
Unconjugated | |
UBE2N | |
BLU, UBC13, UbcH ben, UBE2N, Ubiquitin carrier protein N, Ubiquitin protein ligase N | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116725, 7334, 93765 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61088, P61089, Q9EQX9 | |
UBE2N | |
UBC13 Fusion Protein Ag0292 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |