missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Ultimus Bottle Top Dispenser
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 30 mL
5 to 60 mL
Increments:
0.05 mL
0.1 mL
0.2 mL
0.5 mL
1.0 mL
2.0 mL
Accuracy:
0.6% (+/-0.015)
0.6% (+/-0.030)
0.6% (+/-0.0360)
0.6% (+/-0.060)
0.6% (+/-0.180)
0.6% (+/-0.600)
Model:
ULT-10
ULT-100
ULT-2.5
ULT-30
ULT-5
ULT-60
Unit Size:
Each
Beschreibung
- With dual inlet technology
- Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
- Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
- Specially designed adaptor allows 360° rotation providing full flexibility
- PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
- Adaptors to fit most of the laboratory reagent bottles
- Colied extension tube for refilling
- Calibration tool provided for convenient and quick in-lab user re-calibration
Specifications:
- Piston: PTFE and ETP
- Valve housing: PFA
- Re-Circulation valve housing: PFA
- Valve assembly: Borosilicate glass ball and seat
- Discharge assembly: PTFE
- Delivery and Inlet tube: FEP
- Compatibility: With all reagents except HF
Spezifikation
Spezifikation
| Accuracy | 0.6% (+/-0.015) |
| Product Type | Bottletop Dispenser |
| Certifications/Compliance | ISO 8655 standards |
| Material | Borosilicate Glass |
| Volume (Metric) | 0.25 to 2.5 mL |
| Increments | 0.05 mL |
| Model | ULT-2.5 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur