Learn More
Thermo Scientific™ Ultra Clear qPCR Caps, strips of 8, Sustain™ Series
Thermo Scientific™ Ultra Clear qPCR Caps, strips of 8, Sustain™ Series, are designed to work with PCR plastics and Sustain Series standard low profile strip tubes and 96-well plates.
Brand:  Thermo Scientific™ AB0866SS
37.66 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 30179681
  • 56.50 € / Pack of 120
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
Thermo Scientific™ Ultra Clear qPCR Caps, strips of 8, Sustain™ Series, are designed to work with PCR plastics and Sustain Series standard low profile strip tubes and 96-well plates. The strip caps are easy to apply and create a secure seal to prevent minimize evaporation.
Features and benefits
  • Suitable for qPCR and PCR applications
  • Flat surface of each cap facilitates sample labeling
  • Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
Polypropylene
120 Strips
Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Autoclavable
Less waste, renewable material