Learn More
OHAUS™ Ultra Multi-Purpose Clamps
OHAUS™ Ultra Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30392251
 View more versions of this product
Product Code. 15752299
  • 56.40 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in nickel-plated zinc or stainless steel
  • Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multi-purpose Clamp
Non-slip vinyl sleeves and fiberglass covers
11 mm
5.000 in.
127 mm
0 to 70 mm
Nickel-plated
Clamp, Multi Purpose, CLM-ULTRA3DZM
8.74 in.
222.00 mm
Medium
3-Prong Dual Adjust