Learn More
UVP Ultra Violet EL Lamps
Brand:  UVP 95-0250-02
Product Code. 11758251
  • 851.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
UV Lamp
230 V
302 nm
376 x 97 x 64 mm
64 mm
2 UV Tubes
1 Ea.
UV-B
8 W
0.32 A
8 W, 230 V, 50/60 Hz
50/60 Hz
376 mm
97 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products