missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureSpin™ 632 Swing-out Ultracentrifuge Rotor

Performance for purification applications and flexibility of additional bucket options on one ultracentrifuge rotor.

Brand:  Thermo Scientific™ 75003032

 View more versions of this product

Product Code. 16479639

  • 36481.50 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Designed for purification applications, the Thermo Scientific™ SureSpin™ 632 swing-out ultracentrifuge rotor features top loading design for ease-of-use and the versatility of 38.5mL or additional bucket set option for 17mL bucket capacity. Available for Thermo Scientific™ Sorvall™ WX+ ultracentrifuges.

Features
  • Top loading design for gentle treatment of delicate gradients provides ease-of-use, while mitigating mishaps from a mishooked bucket
  • Flexibility of additional 17mL bucket set to provide double the versatility with one rotor
  • Utilizes the same tubes and adapters as the Thermo Scientific AH-629 and SureSpin 630 rotors  for seamless transition to the enhanced SureSpin 632 rotors
  • Fully integrated for use with Sorvall WX+ ultracentrifuges
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SureSpin 632 Rotor (38.5mL)
Swinging Bucket Rotor
187,295 x g
16.36 cm
194
2.5 cm
Sorvall WX+ ultraCentrifuge
Titanium
6 x 38.5 mL
32,000 rpm
7.46 cm
8.9 cm
15.87 lb. (7.2 kg)
Buckets, rack, rotor stand, thread lubricant, o-rings, o-ring grease, extra speed disk (32,000rpm), rotor log book and instruction manual
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.