Learn More
Thermo Scientific™ UltraFast™ UFC-WAX GC Columns
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns.
2433.72 € - 3040.49 €
Specifications
For Use With (Application) GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances
Length (Metric) 5 m
Diameter (Metric) Inner 0.1 mm
Stationary Phase UltraFast UFC-WAX
For Use With (Equipment) Trace GC UltraFast Instrument
View More Specs
Products 2
Product Code Brand Film Thickness Price Quantity & Availability  
Product Code Brand Film Thickness Price Quantity & Availability  
10594265
View Documents
Thermo Scientific™
UFMC00001010501
0.1 μm N/A N/A N/A
10137934
View Documents
Thermo Scientific™
UFMC00001010503
0.2 μm N/A N/A N/A
Description
Show More
Specifications
Specifications
GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances
0.1 mm
Trace GC UltraFast Instrument
5 m
UltraFast UFC-WAX
Ultrafast
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ UltraFast™ UFC-WAX GC Columns