Learn More
Merck Ultrafree™-CL Centrifugal Filters
For use with protein and nucleic acid solutions
344.00 €
Specifications
Disposable Single-use
Material (Membrane) PTFE (Hydrophilic)
Sterility Non-sterile
Housing Material PTFE
Quantity 25Pack
View More Specs
Products 1
Product Code Brand Color Type Price Quantity & Availability  
Product Code Brand Color Type Price Quantity & Availability  
10298561
View Documents Promotion Details
Merck
UFC40LG25
Yellow Centrifugal Filters
344.00 €
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
Ultrafree™-MC (0.5 mL) and Ultrafree™-CL (2 mL) centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes (from 0.1 to 5.0 μm) for fast filtration and highly reproducible performance. Pre-sterilized units are also available. Use in fixed-angle rotors for 1.5 mL tubes (-MC) and 15 mL tubes (–CL).•Five different pore sizes from 0.1 to 5.0 μm •Pre-sterilized units also available •Fast filtration and highly reproducible performance •Use in fixed-angle rotors for 1.5 mL tubes Applications•Removal of particles and precipitates from aqueous and some solvent-based samples •Ideal for use with protein and nucleic acid solutions
Specifications
Specifications
Single-use
Non-sterile
25Pack
0.4 mL
16.3 mm
PTFE (Hydrophilic)
PTFE
Centrifugal Filters
77 cm