Learn More
Vileda Professional™ UltraSpeed Pro MicroStrong Controlled Environments Mop
The UltraSpeed Pro MicroStrong Mop is a durable, 100% polyethersulfone microfiber looped mop for use with the UltraSpeed Pro CE System.
Brand:  Vileda Professional™ 157778
Product Code. 17852496
  • 380.00 € / Pack of 20
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reusable, single-sided microfiber mop
  • 40 cm including mop label with LOT code
  • Usable up to ISO class 4 to 9 (GMP A/B, C, D)
  • Strong and durable loop microfiber construction
  • High absorbency
  • Low particle shedding
Specifications
Specifications
Mop Pad
Non-sterile
Reusable
Polyester
White
Not Autoclavable
No