Learn More
Epredia™ UltraVision™ Quanto Detection System HRP DAB
Epredia™ UltraVision Quanto HRP DAB delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.
Brand: Epredia™
Description
UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.- Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
- Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
- Reduced cost per test — Quanto remains one of the most affordable systems available
- Shorter incubation times — Faster run times
Includes: TL-060-QHD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Primary Antibody Amplifier Quanto(TL-060-QPB), 60mL HRP Polymer Quanto(TL-060-QPH), 60mL Ultra V Block (TA-060-UB), 60mL DAB Quanto Substrate (TA-060-QHSX), 2mL DAB Quanto Chromogen (TA-002-QHCX)
TL-125-QHD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Primary Antibody Amplifier Quanto(TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB), 125mL DAB Quanto Substrate (TA-125-QHSX), 4mL DAB Quanto Chromogen (TA-004-QHCX)
Specifications
60 mL | |
60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Primary Antibody Amplifier Quanto(TL-060-QPB), 60mL HRP Polymer Quanto(TL-060-QPH), 60mL Ultra V Block (TA-060-UB), 60mL DAB Quanto Substrate (TA-060-QHSX), 2mL DAB Quanto Chromogen (TA-002-QHCX) | |
Store at 2-8°C. Each component is stable for up to 18 months. |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.