missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision™ Quanto Mouse on Mouse

Provide strong staining while reducing mouse on mouse background all with a simple routine procedure using the Epredia™ UltraVision™ Quanto Mouse on Mouse HRP IHC detection system.

Brand:  Epredia™ TL-060-QHDM


Product Code. 12674007

  • 1645.00 € / 60mL
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Epredia UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely. UltraVision Quanto Makes Optimal Results Routine by providing:

  • Strong Staining
  • Reduced Mouse on Mouse Background
  • Simple Biotin Free Polymer Procedure
  • For use with a DAB or other HRP Chromogen; For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL)
Specifications

Specifications

200 to 600 tests
60 mL
Store at 2-8°C. Stable for up to 18 months.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 60ML ULTRAVISION QUANTO MOUSE ON MOUSE HRP, FOR use with a DAB or other HRP Chromogen, for optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL) 200 to 600 tests 60mL

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : 2-methyl-3(2H)-isothiazolone, 5-Bromo-5-nitro-1,3-dioxane

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.