Description
- Earplugs with a tri-flange design that helps keep the earplug in place
- Supplied with an attractive storage case featuring a belt clip, making it convenient for use and reducing contamination
- Earplugs are reusable and no roll down required
- Finger grip design improves ease of insertion
- One size fits the majority of wearers
Specifications
Specifications
Color | Orange |
Certifications/Compliance | CE Approved |
Size | Universal |
Pairs per Package | 50 Pairs/Box |
Noise Reduction Rating (NRR) | 25 dB |
Corded | Uncorded |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
3M™ Uncorded Earplugs >
Spot an opportunity for improvement?Share a Content Correction