missing translation for 'onlineSavingsMsg'
Learn More

BICASA Undercounter Drawer Cabinet on Casters

Undercounter Drawer Cabinet on Casters with fire-retardant particleboard body and soft closing drawers and single lock.

Brand:  BICASA TN.DR.0900.760

864.01 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 17427423

  • 984.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Undercounter cabinets on casters
  • Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads)
    • 2 are lockable with brake, Height 100 mm
    • Central locking system as standard
    • Anti-tilt safety system to open only one drawer at a time
  • Body width 500 mm + 20 mm of ergonomic anodized and painted aluminum handle
  • Available Facade Colors: Gray, Red, Blue, Anthracite
Specifications

Specifications

Light Gray
Mobile
900 x 520 x 760 mm
Yes
Not Autoclavable
EN 16121
4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.