Learn More
Haldenwanger™ Glazed Porcelain Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Mortars are constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ L 55/4/RAUH
This item is not returnable.
View return policy
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
250 mL | |
115mm | |
Porcelain |
Unglazed | |
Mortar | |
63mm |