Learn More
Duran™ Ungraduated Measuring Cylinders
These measuring cylinders are for holding and measuring varying amounts of liquids in the laboratory.
Brand:  Duran™ 213913679
 View more versions of this product
Product Code. 15513162
  • 149.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Without graduation
  • Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits
  • Robust, virtually inert and exceptionally resistant to high temperatures
  • Outstanding corrosion resistance
  • Manufactured from borosilicate glass 3.3
  • Large hexagonal base has three knobs to increase stability and prevents cylinder from rolling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laboratories
Ungraduated Measuring Cylinder
Borosilicate Glass
DIN EN ISO 4788
Hexagonal
250 mL
10/pk
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Duran™ Ungraduated Measuring Cylinders > Volume: 250 mL