Learn More
Hughes Unheated Emergency Safety Shower
Floor mounted unheated emergency safety shower, suitable for use in all applications.
566.00 € - 1030.00 €
Specifications
Mounting Floor
Product Type Safety Shower
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
17702358
View Documents
Hughes
HSHS0018GS
Stainless Steel
1030.00 €
Each
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
17712358
View Documents
Hughes
HSHS0018GV
Galvanized Steel
566.00 €
Each
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
Description
Description
  • Floor mounted unheated emergency safety shower
  • ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
  • Delivers the ANSI recommended 76 liters per minute of potable water
  • Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Specifications
Floor
Safety Shower
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower