Learn More
Abeba™ UNI6 1740 Shoes
Product Code. 15785248
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15785248
Brand: Abeba™ 1740/42
Only null left
Add to basket
Add to basket
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color White
Material Microfiber
Size 42
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1740
Toe Material Steel