Learn More
Abeba™ UNI6 6761 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6761/39
 View more versions of this product
Product Code. 15692128
  • 127.00 € / 1 pair
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Functional leather with honeycomb pattern
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Leather
Slip-resistant
6761
Black
39
Polyurethane
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ UNI6 6761 Shoes > Size: 39