Learn More
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD
Anthracite trainers made of black water repellent and breathable knitted fabric, slip resistant PU/TPU outsole, steel toe cap
Brand:  ABEBA™ 31775/36
 View more versions of this product
Product Code. 17181185
  • 120.00 € / 1 pair
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made for comfort and safety
  • Comply to EN ISO 20345: 2012, S2, SRC
  • ESD dissipative according EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Specifications
Anthracite
36
ESD according 61340 class climatic 1, EN ISO 20345: 2012 S2 SRC
Trainers
S2 SRC ESD
Knitted Fabric
Steel
Unisex
Water Repellent