Learn More
ABEBA™ Uni6 Black Safety Trainers S3
ABEBA™ Uni6 Black Safety Trainers S3 are made of black tear and water resistant fabric with a slip resistant PU/TPU outsole.
Brand:  ABEBA™ 31628/47
 View more versions of this product
Product Code. 16347981
  • 123.00 € / 1 pair
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfort, safety and style
  • Comply to EN ISO 20345 : 2012, S3, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Tear and water resistant fabric
  • Steel toe cap (200 joules)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable insole Active comfort
Specifications
Specifications
CE EN ISO 20345:2012 S3 SRC ESD
Black
Breathable silver ions lining
Textile
Slip and abrasion resistant, ESD dissipative
Trainer
1 Pair
Water
Laces
Steel
Padded
47
Polyurethane, Thermoplastic Polyurethane
Steel
EN ISO 20345 : 2012
Yes