Learn More
ABEBA™ Uni6 White Safety Slip-On Shoe S2
ABEBA™ Uni6 White Safety Slip-On Shoe S2 made of water and chemical resistant microfiber with a slip resistant PU/TPU outsole.
Brand:  ABEBA™ 31740/47
 View more versions of this product
Product Code. 16328011
  • 112.00 € / 1 pair
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfort, safety and hygiene
  • Comply to EN ISO 20345 : 2012, S2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Water and chemical resistant microfiber
  • Steel toe cap (200 joules)
  • Instep area with elastic band
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • No side stitching
  • Removable insole Active comfort
Specifications
Specifications
CE EN ISO 20345:2012 S2 SRC ESD
BreathaBle silver ions lining
Microfiber
Slip and abrasion resistant, ESD dissipative
Slip-on shoes
1 Pair
Water
White
Padded
47
Polyurethane, Thermoplastic Polyurethane
Steel
EN ISO 20345 : 2012; DIN EN 61340
Yes