Learn More
GE Healthcare Unicorn™ Start 1.0
Control software designed for AKTA™ start. GE Healthcare Unicorn™ Start 1.0 software maximizes the performance of the instrument.
Brand:  GE Healthcare 29018751
Product Code. 15279784
Description
Description
  • An easy to learn and easy to use system to eliminate the hassles of manual protein purification
  • Gain insight from real-time monitoring
  • Your purified proteins can be collected automatically using the Frac30 Purify tagged proteins and antibodies easily - Filtration of protein solutions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Unicorn 1.0
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
GE Healthcare Unicorn™ Start 1.0 > Unicorn 1.0