Learn More
Cytiva (Formerly GE Healthcare Life Sciences) UNICORN start
Control software designed for ÄKTA start
Brand:  Cytiva (Formerly GE Healthcare Life Sciences) 29480207
Product Code. 17059203
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Simple and flexible method creation using predefined templates and phases (steps)
  • Enables real-time monitoring of the run and intuitive way to control the run
  • Evaluate, compare, and share results
  • Create and print PDF reports
  • Robust database storage with easy access to data, data security, and data integrity
  • Maximizes performance
  • Includes:
    • Four different modules:
      • System control
      • Method editor
      • Evaluation
      • Administration
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Control Software
    DVD without license
    Windows™ 7 Professional SP1 (US English 32-Bit and 64-Bit), Windows™ 10 Professional (US English 32-Bit and 64-Bit)
    4 modules