Learn More
Thermo Scientific™ Uniguard™ Direct-Connection Guard Cartridge Holders
Eliminate the requirement for extra fittings with Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders.
Brand:  Thermo Scientific™ 850-WT
77.95 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.
 View more versions of this product
Product Code. 10610781
  • 83.70 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminate the requirement for extra fittings using Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders. They are reusable, stainless-steel guard cartridge holders that attach directly to the analytical column inlet.
  • With PEEK 0.0625in. male outlet that fits many columns
  • 0.0625in. female inlet tip can be used with various standard fittings
Ordering Information:
Other column dimensions are available. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Guard Cartridge Holder for Waters Columns Replacement Tips
Waters Columns Replacement Tips
Cartridge Holder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.