missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unilite 6605 Safety Gloves

Product Code. 17220451
Click to view available options
Size:
6
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight polyamide liner provides mechanical abrasion resistance, while nitrile foam coating is well suited for delicate assembly work.

  • Lightweight, comfortable knit gloves suited for delicate assembly work
  • Breathable nitrile foam coating is ideal for precision mechanical work
  • Enhanced grip in dry or slightly damp areas
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Nitrile Foam
Color Black
Material Nylon
Size 6
EN388 Standard 4122X
For Use With (Application) Inspection, Sorting, and Small Parts Assembly
Gender Unisex
Glove Cuff Style Knit
Series Unilite 6605
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'