missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unilite Thermo HD Safety Gloves

Product Code. 17210491
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Two-layer acylic and cotton terry construction provides protection in cold environment applications. PVC coating on the palm and back of hand enhances grip capabilities, and remains flexible even at low temperatures.

  • Two-layer acrylic, cotton terry contruction provides reliable protection in cold applications
  • PVC coating on the palm and fingers enhances grip capabilites
  • Gloves remain comfortable and flexible, even at low temperatures
  • Provides thermal insulation when in direct contact with cold objects
  • Mechanical protection for heavy-duty tasks
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 511: 12X
Applications Industrial
Coating Material PVC
Color Orange, Black
Material Acrylic, Terrycloth
Size 8
EN388 Standard 3231X
For Use With (Application) Cold Environments, Construction, Forklift Operation, and Refrigeration
Gender Unisex
Glove Cuff Style Knit
Series Unilite Thermo HD
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'