missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unipur 6631 Safety Gloves

Product Code. 17250491
Click to view available options
Size:
5
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyamide gloves with polyurethane coating on the palm and fingertips provide mechanical robustness and abrasion resistance.

  • Lightweight, reliable polyamide safety gloves
  • Polyurethane coating on the inside of the hand and fingertips
  • Ideal for precision assembly work
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Polyurethane
Color Gray
Material Polyamide
Size 5
EN388 Standard 4141X
Gender Unisex
Glove Cuff Style Knit
Series Unipur 6631
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'