Learn More
Sampling Systems™ Unit Dose Sleeve Sampler Set
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Unit Dose Sleeve Sampler Set is ideal for use with a wide range of free flowing powders and granules.
Brand:  Sampling Systems™ 1500S-01
Product Code. 15869261
  • 1505.00 € / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For sampling small volume powders and granules
  • Chemically resistant 316L stainless steel construction
  • Different sample sizes can be taken using the same sampler
  • Includes the following:
    • 1 T bar Handle
    • 4 Handle Extension Rods
    • 1 Type 1 Sampler Chamber
    • 1 Type 2 Sample Chamber
    • 10 Sampling Tips
    • 1 High Quanity Sampler Case
  • All items included are available to be purchased individually
Specifications
Specifications
Unit Dose Sampler Set
Stainless Steel (316L)
Handle, extension rod, sample chambers, sampling tips, and sampler case