Learn More
VELP Scientifica™ Universal H-Stand
The Universal H-Stand ensures stability and compatibility with all VELP™ Overhead Stirrers.
Brand: VELP Scientifica™ A00000366
261.41 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
This item is not returnable.
View return policy
Description
- Great solution for using overhead stirrers on laboratory benches
- Maximum stability thanks to H-shape of the base
- Maximum resistance with premium materials such as aluminum and stainless steel, properly treated to resist chemicals and corrosion
- Possible to adjust stand in order to adapt it to uneven surface
Specifications
Stirrer Stand | |
462 x 430 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
VELP Scientifica™ Universal H-Stand > Universal H-Stand; Dimensions (LxWxH): 462 x 430 mm; Type: Stirrer Stand
Spot an opportunity for improvement?Share a Content Correction