missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Universal Indicator Solution Ph, Honeywell Fluka™
pH 3-10
47.63 € - 127.67 €
Specifications
Chemical Name or Material | Universal Indicator Solution Ph |
---|---|
MDL Number | MFCD00148741 |
pH | 3 to 10 |
Flash Point | 19°C (66.2°F) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15683370
|
Honeywell-Fluka
31282-100ML |
100 mL |
47.63 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603380
|
Honeywell Fluka
31282-6X100ML |
6 x 100 mL | N/A | ||||||
15693370
|
Honeywell-Fluka
31282-1L |
1 L | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Universal Indicator Solution Ph | |
MFCD00148741 | |
3 to 10 | |
19°C (66.2°F) |
Safety and Handling
P210-P280-P305 + P351 + P338-P337 + P313-P403 + P235
H225-H319
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Universal indicator solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title