Learn More
Heidolph Instruments Perforated Platform 1000
For use with Vibramax 100 and Rotamax 120
Brand:  Heidolph Instruments 549-59100-00
Product Code. 10322702
  • 87.15 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Universal perforated platform allows for individual arrangement of vessels
Specifications
Specifications
2 kg
0 to 1100 rpm
Perforated Platform
22 x 22 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products