missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | pH Range | Format | Length (Metric) | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Format | Length (Metric) | Quantity | Price | Quantity & Availability | |||||
11517392
|
Fisherbrand™
0911R00530 |
0.5 to 5.5 | Roll | 5 m | 4 Rolls | N/A | ||||||
11793554
|
Fisherbrand™
0911R00030 |
0.5 to 5 | Roll | 5 m | 1 Roll | N/A | ||||||
12375368
|
Fisherbrand™
0911S00530 |
0.5 to 5 | Tube | 50 mm | 10 Pack | N/A | ||||||
15585669
|
Fisherbrand™
PH111R |
1 to 11 | Roll | 5 m | 1 Roll | N/A | ||||||
12345378
|
Fisherbrand™
0911S00524 |
1 to 11 | Tube | 50 mm | 10 Pack |
61.85 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15505679
|
Fisherbrand™
PH114R |
1 to 14 | Roll | 5 m | 1 Roll | N/A | ||||||
15515679
|
Fisherbrand™
PH114R/4 |
1 to 14 | Roll | 5 m | 4 Rolls | N/A | ||||||
11527392
|
Fisherbrand™
0911R00538 |
4 to 7 | Roll | 5 m | 4 Rolls | N/A | ||||||
12385368
|
Fisherbrand™
0911S00531 |
5 to 9 | Tube | 50 mm | 10 Pack | N/A | ||||||
12395368
|
Fisherbrand™
0911S00532 |
6.5 to 10 | Tube | 50 mm | 10 Pack | N/A | ||||||
11557392
|
Fisherbrand™
0911R00533 |
9 to 13 | Roll | 5 m | 4 Rolls | N/A | ||||||
11713564
|
Fisherbrand™
0911R00033 |
9 to 13 | Roll | 5 m | 1 Roll | N/A | ||||||
12305378
|
Fisherbrand™
0911S00533 |
9 to 13 | Tube | 50 mm | 10 Pack | N/A | ||||||
Description
- Provides rapid and precise readings
- Includes color scale printed on cover for convenience
Applications:
- Used for general analysis in chemical, school laboratories
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Universal pH Indicator Paper