Learn More
Fisherbrand™ Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
4.74 € - 5.95 €
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
11703564
|
Fisherbrand™
0911R00031 |
5 to 9 |
5.95 €
Each |
In Stock
Log in to see stock. |
|||||
11783554
|
Fisherbrand™
0911R00034 |
0 to 10 |
5.90 €
Each |
In Stock
Log in to see stock. |
|||||
11763554
|
Fisherbrand™
0911R00035 |
1 to 14 |
4.74 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Filter paper soaked with one or several colored indicators of pH
- Economic range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2