Learn More
Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Brand:  Thermo Scientific™ 236019
 View more versions of this product
Product Code. 10462902
  • 286.43 € / Each
Estimated Shipment: 28-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2800 mL
Flask Clamp
2800mL Fernbach Flask Clamp
MaxQ™ 8000 Shaker
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products