Learn More
Grant Instruments™ Universal Tray for Grant Shaking Water Baths
Holds flasks, bottles, and other vessels in place when used with Grant Shaking Water Baths. Includes adjustable springs.
Brand:  Grant Instruments™ TU12
 View more versions of this product
Product Code. 15315812
  • 335.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Holds flasks, bottles, and other vessels in place when used with Grant Shaking Water Baths. Includes adjustable springs.
  • Universal tray with adjustable springs
  • Highly versatile for a variety of vessel types
Specifications
Specifications
LSB12
Universal Tray