missing translation for 'onlineSavingsMsg'
Learn More

Universol- ES liquid scintillation cocktail, MP Biomedicals™

307.00 € - 872.00 €

Specifications

Chemical Name or Material Universol- ES liquid scintillation cocktail
Density 0.9 to 1.0g/mL at 20°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15762252
View Documents
MP Biomedicals
0188248001
1 gal.
307.00 €
3.79 L
Please to purchase this item. Need a web account? Register with us today!
15772252
View Documents
MP Biomedicals
0188248004
4 x 1 gal.
872.00 €
15.14 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Universol- ES liquid scintillation cocktail
0.9 to 1.0g/mL at 20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Universol ES

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethoxylated nonylphenol, PXE, 2-methoxyethanol

SDS
Documents

Documents

Product Certifications