Learn More
Macherey-Nagel™ Unmodified Cellulose MN 2100
Brand: Macherey-Nagel™ 815060.5
This item is not returnable.
View return policy
Specifications
Purified | |
5 kg | |
∼5500 cm2/g | |
Unmodified Cellulose MN 2100 |