missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ L-Shaped Untapered Connectors

Designed for use with hose system and vacuum lines.

Brand:  Kartell™ 0125300

 View more versions of this product

Product Code. 12513297

  • 48.70 € / Pack of 100
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions

Compliance

Meet Food and Agriculture regulations

Specifications

Specifications

Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Autoclavable
Useful for any hoses system and vacuum line
6mm bore
Untapered L Tubing Connector
10 mm
Polypropylene
100/Pk.
L-shaped
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.