missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human uPa/PRAP1 ELISA Kit
ELISA
Brand: Invitrogen™ EHPRAP1X10
This item is not returnable.
View return policy
Description
Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.
Specifications
Q96NZ9 | |
10 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
PRAP1 | |
<10 | |
HRP | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
13.11 to 3200 pg/mL | |
HRP | |
Plasma, Serum, Supernatant | |
118471 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
PRAP1 | |
4 hrs 45 mins |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction