missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Flangeless Fittings: Ferrules, Tefzel
No need to flange tubing
Brand: Kinesis™ P-359X
This item is not returnable.
View return policy
Description
No need to flange tubing
Excellent for connecting rigid or semi-rigid tubingSpecifications
Flangeless Ferrule | |
0.2 in. | |
Yellow |
0.16 in. | |
ETFE | |
1/8 in. O.D. Tubing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction