Learn More
Kinesis™ LiteTouch™ Ferrule System; 1/8 in. PEEK Nut
Secure a leak-tight connection
Brand:  Kinesis™ 01939-46
Product Code. 13044160
  • 58.80 € / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Nut
10 to 32 UNF
PEEK
Fits 1/8 in. O.D. tubing
LiteTouch Ferrule System