Learn More
Idex ZDV Low-Pressure Delrin Acetal Unions
Make solid connections in low-pressure applications
7.55 €
Description
- Unions do not include fittings—provide your own to match your tubing size
- Manufactured from inert polymers
- True ZDV union
- Acetal body
Specifications
Union | |
60°C | |
Acetal |