Learn More
Kinesis™ ZDW Tefzel Low-Pressure Unions
Make solid connections in low-pressure applications
Brand: Kinesis™ 02023-47
This item is not returnable.
View return policy
Description
Make solid connections in low-pressure applications
- Unions do not include fittings—provide your own to match your tubing size
- Manufactured from inert polymers
- True ZDV union
- ETFE body
Specifications
Union | |
ETFE | |
1/8 in. OD Tubing |
80°C | |
Natural |